Thermo Scientific™

Pierce™ Fluorescence Biotin Quantitation Kit

Catalog number: 46610
Have Questions?
Thermo Scientific™

Pierce™ Fluorescence Biotin Quantitation Kit

Catalog number: 46610
Have Questions?
Catalog Number
46610
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
466101 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Fluorescence Biotin Quantitation Kit requires only ten microliters of sample to accurately measure the biotinylation level of labeled antibodies and other biomolecules.

Features of the Fluorescence Biotin Quantitation Kit:

Fast—requires only 5 minute incubation
Economical—requires only 10 μL of sample (approx. 750ng of biotinylated IgG)
Sensitive—linear working range of 10 to 60 picomoles of biotin

This microplate-based biotin assay is easy to perform by adding the supplied fluorescent reporter to the biotinylated samples and diluted biocytin standards. The avidin fluoresces when the weakly interacting HABA (4'-hydroxyazobenzene-2-carboxylic acid) is displaced by the biotin. The amount of biotin is determined by comparing the sample's fluorescence to the biocytin standard curve. This assay requires must less sample volume than the microplate colorimetric HABA assay and is much more sensitive.

Includes
Assay buffer, HABA-avidin fluorescent reporter solution, and biocytin control

Requires
Black opaque 96-well microplate and a fluorescence plate reader (excitation 494 nm; emission 520 nm)

The highly specific interaction of avidin or streptavidin with biotin is the basis for many purification and detection systems. Quantifying the degree of biotinylation is necessary for assessing if a biotin-labeling procedure was successful and what amount of biotinylated molecule to use for a specific application.

HABA (4'-hydroxyazobenzene-2-carboxylic acid) is a dye that weakly interacts with avidin and is commonly used in a colorimetric assay to quickly estimate the biotin-to-protein ratio. The Fluorescence Biotin Quantitation Kit also uses HABA dye but is more sensitive and accurate. A premix of fluorescent avidin with HABA (DyLight Reporter) is added to the solution containing the biotinylated sample. Because of its higher affinity for avidin, biotin displaces the HABA, allowing the avidin to fluoresce. The amount of biotin is measured in a microplate by comparing the fluorescence to a biocytin standard curve.

The Fluorescence Biotin Quantitation Kit is requires only 10 μL of sample. By contrast, the traditional colorimetric method requires 100 μL of sample, and another leading supplier's fluorescent assay kit requires 50 μL of sample. Our kit is more accurate than the other supplier's fluorescent biotin assay (Brand I).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Fluorescence Biotin Quantitation Kit
Product Line
Pierce™
Label or Dye
Biotin
Labeling Target
Antibodies, Proteins

Contents & Storage

Sufficient for 216 samples in microplate format

• Phosphate Buffered Saline (20X), 3 mL
• DyLight Reporter, 1.3 mL
• 1 mM Biocytin Control, 0.1 mL

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text