Finnpipette™ F1 Multichannel Pipettes
Finnpipette™ F1 Multichannel Pipettes
Thermo Scientific™

Finnpipette™ F1 Multichannel Pipettes

Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of ChannelsColor Code
4661010N5 to 50 μL8Yellow
4661000N1 to 10 μL8Pink
4661020N10 to 100 μL8Yellow
4661030N30 to 300 μL8Orange
4661040N1 to 10 μL12Pink
4661050N5 to 50 μL12Yellow
4661060N10 to 100 μL12Yellow
4661070N30 to 300 μL12Orange
4661080N1 to 10 μL16Purple
4661090N5 to 50 μL16Turquoise
Catalog number 4661010N
Price (USD)/ Each
1,017.00
Online exclusive
1,130.00 
Save 113.00 (10%)
-
Add to cart
Volume (Metric):
5 to 50 μL
Number of Channels:
8
Color Code:
Yellow
Price (USD)/ Each
1,017.00
Online exclusive
1,130.00 
Save 113.00 (10%)
Add to cart
Finnpipette™ F1 Multichannel Pipettes
Catalog number4661010N
Price (USD)/ Each
1,017.00
Online exclusive
1,130.00 
Save 113.00 (10%)
-
Add to cart

Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F1: Everything you want in your everyday, go-to pipette

Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.

Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.

5 Year Warranty
With convenient online registration, and may vary by country.

Learn more about the complete Finnpipette family >

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette F1
ColorYellow
Number of Channels8
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach