Finnpipette™ F1 Multichannel Pipettes
Finnpipette™ F1 Multichannel Pipettes
Thermo Scientific™

Finnpipette™ F1 Multichannel Pipettes

Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of ChannelsColor Code
4661030N30 to 300 μL8Orange
4661000N1 to 10 μL8Pink
4661010N5 to 50 μL8Yellow
4661020N10 to 100 μL8Yellow
4661040N1 to 10 μL12Pink
4661050N5 to 50 μL12Yellow
4661060N10 to 100 μL12Yellow
4661070N30 to 300 μL12Orange
4661080N1 to 10 μL16Purple
4661090N5 to 50 μL16Turquoise
Catalog number 4661030N
Price (USD)
1,017.00
Online Exclusive
1,153.00
Save 136.00 (12%)
Each
Add to cart
Volume (Metric):
30 to 300 μL
Number of Channels:
8
Color Code:
Orange
Price (USD)
1,017.00
Online Exclusive
1,153.00
Save 136.00 (12%)
Each
Add to cart
Ask our AI about this Product

Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F1: Everything you want in your everyday, go-to pipette

Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.

Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.

5 Year Warranty
With convenient online registration, and may vary by country.

Learn more about the complete Finnpipette family >

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F1
Increments1μL
ColorOrange
Color CodeOrange
Number of Channels8
Volume (Metric)30 to 300 μL
Unit SizeEach

Additional Finnpipette F1 Models

Finnpipette Novus Electronic Pipette Models

Finntip Pipette Tips

Interested in NEW ClipTip Technology?

Pipette Service

Calibration and Preventative Maintenance

Whether it's cGLP, cGMP, critical research, academia, or special requirements; our experienced service personnel are highly trained in providing exceptional care for all pipette models. Our Unity Lab Services team offers three levels of service for pipettes.

Request a Pipette Service Return Kit (North America Only) ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection