Polydextrose
Polydextrose
Polydextrose
Thermo Scientific Chemicals

Polydextrose

CAS: 68424-04-4 | (C6H10O5)n
Have Questions?
Catalog NumberQuantity
4663600505 g
Catalog number 466360050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS68424-04-4
IUPAC NamePolydextrose
Molecular Formula(C6H10O5)n
SMILESOCC1OC(-*)[C@@H](O)C(O)[C@@H]1O-*
MDL NumberMFCD00804499
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellowisch
Appearance (Form)Powder
Loss on drying=<4 %
Purity>=88 % (polymer)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468130Certificate of AnalysisJan 25, 2025466360250, 466360050
A0451243Certificate of AnalysisMay 02, 2023466360250, 466360050
A0417561Certificate of AnalysisSep 26, 2022466360250, 466360050
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection