Thermo Scientific Chemicals

Polydextrose, Thermo Scientific Chemicals

Catalog number: 466360250
25 g, Each
Thermo Scientific Chemicals

Polydextrose, Thermo Scientific Chemicals

Catalog number: 466360250
25 g, Each
Quantity
Catalog number: 466360250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68424-04-4
IUPAC Name
Polydextrose
Molecular Formula
(C6H10O5)n
SMILES
OCC1OC(-*)[C@@H](O)C(O)[C@@H]1O-*
MDL Number
MFCD00804499
Appearance (Color)
White to yellowisch
Appearance (Form)
Powder
Loss on drying
=<4 %
Purity
>=88 % (polymer)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text