Thermo Scientific™

SuperSignal™ Western Blot Enhancer

Catalog number: 46641
Have Questions?
Thermo Scientific™

SuperSignal™ Western Blot Enhancer

Catalog number: 46641
Have Questions?
Catalog Number
46641
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4664150 mL
-
46640500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to a detection performed without it.

Features of SuperSignal Western Blot Enhancer:

Increase sensitivity—achieve 3- to 10-fold increase in signal intensity and sensitivity
Improve specificity—improves signal-to-noise ratio for poor quality and low affinity antibodies
Better clarity—reduces background for cleaner Western blots
Membrane compatibility—provides effective signal enhancement with PVDF and nitrocellulose membranes
Substrate compatibility—validated for use with chromogenic, chemiluminescent and fluorescent detection methods

When a protein or antigen is difficult to detect because of low abundance or poor immunoreactivity, use of SuperSignal Western Blot Enhancer can significantly reduce background and enhance detection of low-abundance and weakly immunoreactive antigens.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Western Blot Enhancer
Detection Method
Chemiluminescent, Fluorescent, Colorimetric
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
SuperSignal™
Quantity
50 mL

Contents & Storage

Upon receipt store solutions at 4°C Products are shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text