Timolol maleate
Thermo Scientific Chemicals

Timolol maleate

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466512500
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
Add to cart
Timolol maleate
Catalog number466512500
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS26921-17-5
IUPAC Name(2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
Molecular FormulaC17H28N4O7S
InChI KeyWLRMANUAADYWEA-NWASOUNVSA-N
SMILESOC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.0 to 102.0 %
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only