Thermo Scientific Chemicals

Dodecenylsuccinic acid, Thermo Scientific Chemicals

Catalog number: 466600010
1 g, Each
Thermo Scientific Chemicals

Dodecenylsuccinic acid, Thermo Scientific Chemicals

Catalog number: 466600010
1 g, Each
Quantity
Catalog number: 466600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29658-97-7
IUPAC Name
2-[(1E)-dodec-1-en-1-yl]butanedioic acid
Molecular Formula
C16H28O4
InChI Key
QDCPNGVVOWVKJG-VAWYXSNFNA-N
SMILES
CCCCCCCCCC\C=C\C(CC(O)=O)C(O)=O
Appearance (Form)
viscous liquid
Acid value
235 to 300 (mg KOH/g)
Appearance (Color)
Clear yellow to red-brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text