Thermo Scientific Chemicals

Genipin

Catalog number: 466640010
1 g, Each
Thermo Scientific Chemicals

Genipin

Catalog number: 466640010
1 g, Each
Quantity
Catalog number: 466640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6902-77-8
IUPAC Name
methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Molecular Formula
C11H14O5
InChI Key
AZKVWQKMDGGDSV-BCMRRPTOSA-N
SMILES
COC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
118.0°C to 123.0°C
HPLC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text