Maltodextrin 12
Thermo Scientific Chemicals

Maltodextrin 12

CAS: 9050-36-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 466680010
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
-
Add to cart
Quantity:
1 kg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS9050-36-6
IUPAC Name(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Molecular FormulaC6H12O6
InChI KeyGZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILESOC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<7.0 %
Appearance (Color)White
Infrared spectrumConforms
Dextrose equivalent9.0 to 15.0
Appearance (Form)powder or granular powder
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0418710Certificate of AnalysisMay 17, 2025466682500, 466680010, 466680500
A0467417Certificate of AnalysisJan 25, 2025466682500, 466680010, 466680500
A0462721Certificate of AnalysisMay 17, 2024466682500, 466680010, 466680500
A0461362Certificate of AnalysisMar 22, 2024466682500, 466680010, 466680500
A0457960Certificate of AnalysisDec 05, 2023466682500, 466680010, 466680500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection