Chemical Identifiers
IUPAC Name(5R,6S)-5-ethenyl-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,5H,6H-pyrano[3,4-c]pyran-1-one
Molecular FormulaC16H20O9
InChI KeyDUAGQYUORDTXOR-GPQRQXLASA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol)356.33
View more