Pyrroloquinoline quinone
Pyrroloquinoline quinone
Thermo Scientific Chemicals

Pyrroloquinoline quinone

CAS: 72909-34-3 | C14H6N2O8, C14H6N2O8 | 330.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467012500
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
-
Add to cart
Quantity:
250 mg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS72909-34-3
IUPAC Name4,5-dioxo-1H,4H,5H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Molecular FormulaC14H6N2O8
InChI KeyMMXZSJMASHPLLR-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=C(N1)C1=C(C=C(N=C1C(=O)C2=O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder
Infrared spectrumConforms
HPLC>=95.0 %
Appearance (Color)Dark orange to red to brown
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464415Certificate of AnalysisSep 06, 2024467012500, 467010010
A0462885Certificate of AnalysisJun 10, 2024467012500, 467010010
A0447588Certificate of AnalysisNov 30, 2022467012500, 467010010
A0443623Certificate of AnalysisJul 27, 2022467012500, 467010010
A0419185Certificate of AnalysisJul 08, 2020467012500, 467010010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link