(+)-trans-Taxifolin
Thermo Scientific Chemicals

(+)-trans-Taxifolin

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 467035000
Price (USD)
602.65
Online Exclusive
669.00
Save 66.35 (10%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
602.65
Online Exclusive
669.00
Save 66.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS480-18-2
IUPAC Name(2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC15H12O7
InChI KeyCXQWRCVTCMQVQX-LSDHHAIUSA-N
SMILESO[C@@H]1[C@H](OC2=CC(O)=CC(O)=C2C1=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<3.0 %
Specific optical rotation+19.0° to +23.0° (20°C, 589 nm) (C=1,MeOH)
HPLC>=98.0 %
Appearance (Color)White to yellow to light brown
Appearance (Form)powder
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection