1x1 image pixel for data collection
Tianeptine, sodium salt
Tianeptine, sodium salt
Thermo Scientific Chemicals

Tianeptine, sodium salt

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467052500
Price (USD)
133.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
133.00
Each
Add to cart
Chemical Identifiers
CAS30123-17-2
IUPAC Namesodium 2-[(6-carboxyhexyl)amino]-6-chloro-10-methyl-9λ⁴-thia-10-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-9,9-bis(olate)
Molecular FormulaC21H25ClN2NaO4S
InChI KeyKBNZQVDKURREAX-UHFFFAOYNA-L
SMILES[Na+].CN1C2=CC=CC=C2C(NCCCCCCC(O)=O)C2=CC=C(Cl)C=C2S1([O-])[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Powder
Melting point183°C to 187°C
Water=<3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection