1x1 image pixel for data collection
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
Thermo Scientific™

E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes

Automate daily 96-and 384-well microplate pipetting tasks with the sealed-in security of ClipTip technology in these electronic multichannel pipettes, which offer both versatility and customization.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of ChannelsCompatible Tips
4671030BT1 to 30 μL16ClipTip 384 30
4671000BT0.5 to 12.5 μL8ClipTip 12.5
4671040BT2 to 125 μL8ClipTip 200
4671070BT10 to 300 μL8ClipTip 300
4671100BT15 to 1250 μL8ClipTip 1250
4671010BT0.5 to 12.5 μL12ClipTip 12.5
4671050BT2 to 125 μL12ClipTip 200
4671080BT10 to 300 μL12ClipTip 300
4671090BT30 to 850 μL12ClipTip 1000
4671020BT0.5 to 12.5 μL16ClipTip 384 12.5
4671060BT2 to 125 μL16ClipTip 384 125
Catalog number 4671030BT
Price (USD)
2,248.65
Online Exclusive
2,438.00
Save 189.35 (8%)
Each
In stock
Add to cart
Volume (Metric):
1 to 30 μL
Number of Channels:
16
Compatible Tips:
ClipTip 384 30
Price (USD)
2,248.65
Online Exclusive
2,438.00
Save 189.35 (8%)
Each
Add to cart

With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed-in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC.

Innovative ClipTip interlocking tip interface technology*
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette

My Pipette Creator to revolutionize the way you pipette with a new era of technology
  • New web-based pipetting app powered by Thermo Fisher Cloud
  • Enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues
  • Offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols
  • Connect E1-ClipTip pipette to the app via a Bluetooth** or USB connection.
  • Accelerate collaboration and reproducibility, all from the comfort of your computer.

E1-ClipTip pipette to help you enjoy pipetting
  • E1-ClipTip is designed for optimum comfort and ease of use: the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
  • Two operating options: Step-based Matrix or Presets with settings for your most common applications.
  • The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name.
  • Connect your E1-ClipTip pipette to My Pipette Creator app conveniently via Bluetooth**
  • Five different user profiles offer customized settings for multiple users and protocols
  • MyPipette™ function to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
  • Service and calibration trackers remind you when the pipette needs to be sent for service
  • Li-ion battery for long lasting performance and fast recharging. Recharging with a charging stand or a plug, you decide.
  • 2-year warranty with convenient web registration

E1-ClipTip pipette and ClipTip tips for every application
  • Choose your E1-ClipTip, volume range from 0.5μL to 1250μL:
    • Single channels
    • Multichannels with 8, 12 or 16 channels in 96 and 384 formats
    • Adjustable tip spacing multichannels with 6, 8, 12 channels in 96 and 384 formats
  • Wide selection of ClipTip tips:
    • ClipTip 96-format tips are available in 20, 50, 200, 300, 1000 and 1250μl
    • ClipTip 384-format tips are available in 12.5, 30 and 125μl
    • Several packaging options: filtered, sterile, non-sterile, racked and reload


* E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology.
** Bluetooth models available in following countries/regions: Andorra, Australia, Austria, Belgium, Bulgaria, Canada, China, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hong Kong, Hungary, Iceland, India, Ireland, Italy, Japan, Korea (Rep. of), Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Monaco, Netherlands, New Zealand, Norway, Poland, Portugal, Romania, San Marino, Singapore, Slovakia, Slovenia, Spain, Sweden, Switzerland, Taiwan, United Kingdom, United States and Vatican. For other countries catalog numbers without 'BT' extension should be used.




All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. The Bluetooth™ is a trademark of Bluetooth SIG, Inc.
Specifications
Compatible TipsClipTip 384 30
Increments0.01μL
ColorViolet
For Use WithClipTip™ 384 30
ModelBluetooth
Number of Channels16
Product LineE1-ClipTip
ProgrammabilityYes
TypeElectronic Pipette
Volume (Metric)1 to 30 μL
Unit SizeEach

Go beyond traditional limitations with My Pipette™ Creator App.

Get connected ›

Have an earlier model E1-Cliptip Electronic Pipette and want to connect to My Pipette Creator?

NOW AVAILABLE: Software Upgrade Kit for E1-ClipTip Pipettes with software versions 1.27 or earlier
Learn more ›

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection