Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.
Breakapart | No |
Binding Property | Slightly Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells |
Certified | Yes |
Format | Strip Plate |
Lid | With Lid |
Detection Method | Colorimetric |
Packaging | Inner packs of 10 |
Binding Type | Adsorption |
Color | Clear |
Product Type | Frame |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Sterility | Non-sterile |
Surface Treatment | MediSorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Proteins |
Well Design | F8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells |
Certified | Yes |
Format | Standard |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 120 Cases |
Binding Type | Adsorption |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Sterility | Non-sterile |
Surface Treatment | MaxiSorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 120 |
No. per Pack | 20 |
Plate Blocking | None |
Quantity | 120/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | F8 |
Unit Size | Case of 120 |
Breakapart | No |
Binding Property | Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells |
Certified | Yes |
Format | Standard |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 60 Cases |
Color | Clear |
Product Type | Frame |
Material | Polystyrene |
Volume (Metric) Well | 380 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | C8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Detection Method | Colorimetric |
Packaging | 60 Cases, 10 Pk. |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
No. of Wells | 8 |
Volume (Metric) Working | 250 μL |
Surface Treatment | PolySorp |
Well Shape | Round, C-bottom |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Lipids |
Well Design | C8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 60 Cases |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Volume (Metric) Working | 250 μL |
Surface Treatment | MaxiSorp |
Well Shape | Round, C-bottom |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | C8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Very Hydrophilic |
Certified | No |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 10 Pk., 60 Cases |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Surface Treatment | MultiSorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Glycans |
Well Design | F8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells. |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 60 Cases |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Sterility | Non-sterile |
Surface Treatment | MaxiSorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | F16 (2x8 wells) |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophobic |
Certified | No |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Colorimetric |
Packaging | 60 Cases, 10 Pk. |
Color | Clear |
Product Type | Frame |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 16 |
Volume (Metric) Working | 350 μL |
Surface Treatment | PolySorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Target Molecule | Lipids |
Well Design | F16 |
Unit Size | Case of 60 |
Binding Property | Hydrophilic |
Certifications/Compliance | Certified for adsorption of IgG; CV of <5% between wells |
Certified | Yes |
Format | Strip Plate |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 12 inner bags of 5 each |
Color | Clear |
Product Type | Strip Plate |
Material | Polystyrene |
Volume (Metric) Well | 400 μL |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Sterility | Non-sterile |
Surface Treatment | MaxiSorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | ROC 5, ROS 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Antibodies (Glycoproteins) |
Well Design | F8 |
Unit Size | Case of 60 |
Breakapart | No |
Binding Property | Hydrophobic |
Certified | No |
Format | Standard |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Packaging | 60 Cases |
Color | Clear |
Product Type | Microplate Strip |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 350 μL |
Surface Treatment | PolySorp |
Well Shape | Round, Flat Bottom (F-well) |
No. per Case | 60 |
No. per Pack | 10 |
Plate Blocking | None |
Quantity | 60/Cs. |
Target Molecule | Lipids |
Well Design | F8 |
Unit Size | Case of 60 |
Catalog Number | Specifications | Unit Size | Well Design | Surface Treatment | Volume (Metric) Working | Well Shape | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
467120 | Case of 60 | F8 | MediSorp | 350 μL | Round, Flat Bottom (F-well) | 568.80 Online price 632.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
434797 | Case of 120 | F8 | MaxiSorp | 350 μL | Round, Flat Bottom (F-well) | 859.00 Online price 1,258.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
441653 | Case of 60 | C8 | MaxiSorp | 250 μL | Round, C-bottom | 647.65 Online price 708.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
444865 | Case of 60 | C8 | PolySorp | 250 μL | Round, C-bottom | 601.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
445101 | Case of 60 | C8 | MaxiSorp | 250 μL | Round, C-bottom | 568.80 Online price 632.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
467140 | Case of 60 | F8 | MultiSorp | 350 μL | Round, Flat Bottom (F-well) | 675.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
467466 | Case of 60 | F16 (2x8 wells) | MaxiSorp | 350 μL | Round, Flat Bottom (F-well) | 632.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
467679 | Case of 60 | F16 | PolySorp | 350 μL | Round, Flat Bottom (F-well) | 632.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
468667 | Case of 60 | F8 | MaxiSorp | 350 μL | Round, Flat Bottom (F-well) | 567.00 Online price 630.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
469078 | Case of 60 | F8 | PolySorp | 350 μL | Round, Flat Bottom (F-well) | 538.20 Online price 598.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Choice of Surface:
Ordering Information:
Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.