E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
Thermo Scientific™

E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes

Thermo Scientific™ E1-ClipTip™ Equalizer Pipette can be conveniently used with any tube, rack, microplate or horizontal gel box due to the adjustable tip spacing

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerVolumen (metrisch)Anzahl KanäleKompatible Spitzen
4672050BT2 bis 125 μl8ClipTip 200
4672090BT15 bis 1250 μL6ClipSpitze 1250
4672080BT10 bis 300 μl8ClipTip 300
4672100BT15 bis 1250 μL8ClipSpitze 1250
4672010BT0,5 bis 12,5 μl8ClipSpitze 384 12,5
4672030BT1 bis 30 μl8Klemmspitze 384 30
4672060BT2 bis 125 μl8ClipSpitze 384 125
4672020BT0,5 bis 12,5 μl12ClipSpitze 384 12,5
Katalognummer 4672050BT
Preis (EUR)
1.564,16
Exklusiv online
1.903,00
Ersparnis 338,84 (18%)
Each
-
Zum Warenkorb hinzufügen
Volumen (metrisch):
2 bis 125 μl
Anzahl Kanäle:
8
Kompatible Spitzen:
ClipTip 200
Preis (EUR)
1.564,16
Exklusiv online
1.903,00
Ersparnis 338,84 (18%)
Each
Zum Warenkorb hinzufügen
Ask our AI about this Product

Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format.The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC

Innovative ClipTip interlocking tip interface technology*
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette

My Pipette Creator to revolutionize the way you pipette with a new era of technology
  • New web-based pipetting app powered by Thermo Fisher Cloud
  • Enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues
  • Offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols
  • Connect E1-ClipTip pipette to the app via a Bluetooth** or USB connection.
  • Accelerate collaboration and reproducibility, all from the comfort of your computer.

E1-ClipTip pipette to help you enjoy pipetting
  • E1-ClipTip is designed for optimum comfort and ease of use: the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
  • Two operating options: Step-based Matrix or Presets with settings for your most common applications.
  • The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name.
  • Connect your E1-ClipTip pipette to My Pipette Creator app conveniently via Bluetooth**
  • Five different user profiles offer customized settings for multiple users and protocols
  • MyPipette™ function to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
  • Service and calibration trackers remind you when the pipette needs to be sent for service
  • Li-ion battery for long lasting performance and fast recharging. Recharging with a charging stand or a plug, you decide.
  • 2-year warranty with convenient web registration

E1-ClipTip™ Equalizer: transferring multiple samples between different labware formats
  • Adjustable tip spacing allows you to set the distance between pipette tips for different types of labware simply by sliding a scale to expand or contract the tip spacing to the desired format:
    • Microcentrifuge tubes to/from 24,-48,-96-and 384- well microplates
    • Test tubes to/from deepwell blocks
    • Microplates and tubes to agarose gels
  • Save up to 90% in time and repetitions compared to use of single channel manual pipettes***

E1-ClipTip pipette and ClipTip tips for every application
  • Choose your E1-ClipTip, volume range from 0.5 μL to 1250μL:
    • Single channels
    • Multichannels with 8, 12 or 16 channels in 96 and 384 formats
    • Adjustable tip spacing multichannels with 6, 8, 12 channels in 96 and 384 formats
  • Wide selection of ClipTip tips:
    • ClipTip 96-format tips are available in 20, 50, 200, 300, 1000 and 1250 μl
    • ClipTip 384-format tips are available in 12.5, 30 and 125 μl
    • Several packaging options: filtered, sterile, non-sterile, racked and reload

Compatible with Thermo Scientific ClipTip pipette tips.

* E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology.
** Bluetooth models available in following countries/regions: Andorra, Australia, Austria, Belgium, Bulgaria, Canada, China, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hong Kong, Hungary, Iceland, India, Ireland, Italy, Japan, Korea (Rep. of), Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Monaco, Netherlands, New Zealand, Norway, Poland, Portugal, Romania, San Marino, Singapore, Slovakia, Slovenia, Spain, Sweden, Switzerland, Taiwan, United Kingdom, United States and Vatican. For other countries catalog numbers without 'BT' extension should be used.
*** Compared to a manual single channel pipette filling a 96-well microplate from microcentrifuge tubes. See Thermo Scientific Smart Note: Is there an easier and more efficient way to transfer liquids between various labware formats, than using traditional handheld pipettes? 2014.
Specifications
Kompatible SpitzenClipTip 200
BeschreibungE1-ClipTip Entzerrer
Schritten0,1 μl
Verstellbarer Spitzenabstand9 bis 14,2 mm
FarbeGelb
Zur Verwendung mitClipTip 200
ModellBluetooth
Anzahl Kanäle8
ProduktlinieE1-ClipTip™
ProgrammierbarJa
TypMulti-Channel Equalizer Pipette
Volumen (metrisch)2 bis 125 μl
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

0 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection