Chemical Identifiers
CAS130018-87-0
IUPAC Name2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Molecular FormulaC21H27Cl3N2O3
InChI KeyPGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILESCl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
View more