Favipiravir
Thermo Scientific Chemicals

Favipiravir

CAS: 259793-96-9 | C5H4FN3O2, C5H4FN3O2 | 157.10 g/mol
Have Questions?
Catalog NumberQuantity
468691000100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 468691000
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS259793-96-9
IUPAC Name6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Molecular FormulaC5H4FN3O2
InChI KeyZCGNOVWYSGBHAU-UHFFFAOYSA-N
SMILESNC(=O)C1=NC(F)=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<2 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link