Search Thermo Fisher Scientific
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
Description | Finnpipette Novus GLP Kit 4 |
Model | Novus |
Number of Channels | 1 |
Product Line | Finnpipette Novus |
Type | GLP Kit |
Volume (Metric) | 10 to 10,000 μL |
Includes | Novus 100, 1000 and 10000µL single channel pipettes, three Novus stands, three sample Finntip racks |
Unit Size | Each |
Description | Finnpipette Novus GLP Kit 1 |
Color | Assorted |
Model | Finnpipette™ Novus |
Number of Channels | 1 |
Product Line | Finnpipette™ Novus |
Type | GLP Kit |
Volume (Metric) | 10 to 1000 μL |
Includes | Novus 100 and 1000µL single channel pipettes, two Novus stands, two sample Finntip racks |
Unit Size | Each |
Description | Finnpipette Novus GLP Kit 2 |
Color | Assorted |
Model | Novus |
Number of Channels | 1 |
Product Line | Finnpipette Novus |
Type | GLP Kit |
Volume (Metric) | 100 to 5000 μL |
Includes | Novus 1000 and 5000µL single channel pipettes, two Novus stands, two sample Finntip racks |
Unit Size | Each |
Description | Finnpipette Novus GLP Kit 3 |
Model | Novus |
Number of Channels | 1 |
Product Line | Finnpipette Novus |
Type | GLP Kit |
Volume (Metric) | 1 to 1000 μL |
Includes | Novus 10micro, 100 and 1000µL single channel pipettes, three Novus stands, three sample Finntip racks |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4701130 | Each | Finnpipette Novus GLP Kit 4 | Novus 100, 1000 and 10000µL single channel pipettes, three Novus stands, three sample Finntip racks | 2,161.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4700470 | Each | Finnpipette Novus GLP Kit 1 | Novus 100 and 1000µL single channel pipettes, two Novus stands, two sample Finntip racks | 923.00 Online price 1,465.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4700490 | Each | Finnpipette Novus GLP Kit 2 | Novus 1000 and 5000µL single channel pipettes, two Novus stands, two sample Finntip racks | 1,465.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4701120 | Each | Finnpipette Novus GLP Kit 3 | Novus 10micro, 100 and 1000µL single channel pipettes, three Novus stands, three sample Finntip racks | 1,560.65 Online price 2,161.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Novus GLP Kit 1 includes:
Novus GLP Kit 2 includes:
Novus GLP Kit 3 includes:
Novus GLP Kit 4 includes: