Thermo Scientific™

F1-ClipTip™ Good Laboratory Pipetting (GLP) Kits

Catalog number: 4701150N
Have Questions?
Thermo Scientific™

F1-ClipTip™ Good Laboratory Pipetting (GLP) Kits

Catalog number: 4701150N
Have Questions?
Support good laboratory pipettingin your lab with the Thermo Scientific™ F1-ClipTip™ GLP Kits, which offer a variety of single channel pipettes or a mix of single channels and a multichannel pipette packaged with compatible ClipTip tips, and a convenient pipette stand. Select the kit that is right for you, and take advantage of the F1-ClipTip performance and lightweight design to maximize your comfort.
 
Catalog Number
4701150N
Unit Size
Each
Description
F1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL
Volume (Metric)
2 to 1000, 30 to 300 μL
Compatible Tips
ClipTip tips: 20, 200, 300 and 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip tips: 20, 200, 300 and 1000
DescriptionF1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL
Number of Channels1
TypeGLP Kit
Volume (Metric)2 to 1000, 30 to 300 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip GLP Kit 1: 1-10μL, 10-100 μL and 100-1000 μL
Volume (Metric)
1 to 1000 μL
Compatible Tips
ClipTip tips: 12.5 Ext, 200 and 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip tips: 12.5 Ext, 200 and 1000
DescriptionF1-ClipTip GLP Kit 1: 1-10μL, 10-100 μL and 100-1000 μL
Number of Channels1
TypeGLP Kit
Volume (Metric)1 to 1000 μL
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionVolume (Metric)Compatible TipsPrice (USD)AvailabilityQuantity
4701150NFull specifications
EachF1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL2 to 1000, 30 to 300 μLClipTip tips: 20, 200, 300 and 1000
-
Compatible TipsClipTip tips: 20, 200, 300 and 1000
DescriptionF1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL
Number of Channels1
TypeGLP Kit
Volume (Metric)2 to 1000, 30 to 300 μL
Unit SizeEach
4701140NFull specifications
EachF1-ClipTip GLP Kit 1: 1-10μL, 10-100 μL and 100-1000 μL1 to 1000 μLClipTip tips: 12.5 Ext, 200 and 1000
-
Compatible TipsClipTip tips: 12.5 Ext, 200 and 1000
DescriptionF1-ClipTip GLP Kit 1: 1-10μL, 10-100 μL and 100-1000 μL
Number of Channels1
TypeGLP Kit
Volume (Metric)1 to 1000 μL
Unit SizeEach
Showing 2 of 2
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.

Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.

Reduce the risk of loose or leaking tips

When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.

Enable consistent and reproducible pipetting from user to user

Higher-quality results and more efficient research.

Learn more >

Ergonomically in-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Silent, effortless volume adjustment
Enjoy the silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.

Secure volume lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.

Adjustable comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.

Wide Selection of ClipTip Pipette Tips
Choose from seven sizes in 96-format: 12.5 Ext, 20, 50, 200, 300, 300 Ext,1000μL

Lifetime warranty
With convenient online registration, and may vary by country.

Kit 1 includes:
- Three Thermo Scientific™ F1-ClipTip pipettes from 1 μL to 1000 μL
-1-10 μL
-10-100 μL
-100-1000 μL
-Grip Strip for each handle to improve grip
-ClipTip tips: 12.5 Ext: 1 x 96; 200: 1 x 96; 1000: 1 x 96
-F-stand
-Good Laboratory Pipetting Guide
-Reagent reservoir demo pack
 
Kit 2 includes:
-Four Thermo Scientific™ F1-ClipTip pipettes from 2 μL to 1000 μL
-2-20 μL
-20-200 μL
-100-1000 μL
-8-ch 30-300 μL
-Grip Strip for each handle to improve grip
-ClipTip tips: 20: 1 x 96; 200: 1 x 96; 300: 1 x 96; 1000: 1 x 96
-F-stand
-Good Laboratory Pipetting Guide
-Reagent reservoir demo pack


Learn more about the complete ClipTip family >
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text