Plain Microscope Slides, 25 x 75 x 1mm, Economy
Product Image
Thermo Scientific™
Plain Microscope Slides, 25 x 75 x 1mm, Economy
Experience easy handling with the Thermo Scientific™ Plain Microscope Slides, which feature finely ground edges.
Have Questions?
Change viewbuttonViewtableView
Catalog Number
500
500TS
also known as 500
1 x 7 in. Custody Seals
Catalog number 500
Price (USD)
-
Request A Quote
Experience easy handling with the Thermo Scientific™ Plain Microscope Slides, which feature finely ground edges. These slides are manufactured to the highest quality standards for use in hematology, microbiology, and urinalysis as well as other general laboratory and research applications. All slides are precleaned for immediate use. The surface of each slide is completely plain without a frosted tab.
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
Specifications
Unit SizeEach
Share catalog number, name or link