For Use With: Model 3948 and 3962 Environmental Chamber/Incubator
Have Questions?
Catalog number 500182
Price (USD)/ Each
-
Request A Quote
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators.
Specifications
DescriptionReinforced Floor with Removable Ramp
For Use With (Equipment)Model 3948 and 3962 Environmental Chamber/Incubator