Hand Dispenser Kit
Hand Dispenser Kit
Thermo Scientificā„¢
Hand Dispenser Kit
Increase lab flexibility with the Thermo Scientificā„¢ Hand Dispenser Kit, compatible with many Thermo Scientificā„¢ Barnsteadā„¢ water purification systems.
Have Questions?
Catalog number 50138221
Price (USD)/ Each
1,125.00
Online exclusive
1,500.00 
Save 375.00 (25%)
-
Add to cart
Description:
Hand Dispenser Kit
Price (USD)/ Each
1,125.00
Online exclusive
1,500.00 
Save 375.00 (25%)
Add to cart
Hand Dispenser Kit
Catalog number50138221
Price (USD)/ Each
1,125.00
Online exclusive
1,500.00 
Save 375.00 (25%)
-
Add to cart
Increase flexibility in the lab with the Thermo Scientificā„¢ Hand Dispenser Kit, compatible with many Thermo Scientificā„¢ Barnsteadā„¢ water purification systems. This kit includes a dispenser with 3 meter cord that connects to the water system tank and a 0.2 micron final filter. It is compatible with Barnstead LabTowerā„¢, Pacificā„¢ and Smart2Pureā„¢ systems.
Optional for the following Barnstead water purification systems:
  • LabTowerā„¢ EDI
  • LabTowerā„¢ RO
  • LabTowerā„¢ TII
  • Smart2Pureā„¢
  • Pacificā„¢ TII + tank (tank must have optional pump)
  • Pacificā„¢ RO + tank (tank must have optional pump)
Specifications
DescriptionHand Dispenser Kit
For Use With (Equipment)LabTower EDI, LabTower TII, Smart2Pure, Pacific TII + Tank (Tank Must Have Optional Pump), Pacific RO + Tank (Tank Must Have Optional Pump), LabTower RO
IncludesHand Dispenser Kit (includes hand dispenser with 3 meter cord, and 0.2 micron final filter)
Unit SizeEach
1x1 image pixel for data collection