CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving System  for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving System  for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving System  for G-Rex 500M-CS Bioreactors
Thermo Scientific™
CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripciónPara utilizar con (equipo)IncluyeN.º de estantes
50164781Shelving system to support up to 10 G-Rex 500M-CS bioreactors for Heracell VIOS 250i (CR), Forma Steri-Cycle i250 (CR) CO2 incubatorHeracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series3 expanded shelves with reinforced design, shelf supports and shelf rails3
332395G08Shelving system to support G-Rex™ 500M for Reach-In 821L CO2 IncubatorReach-In 821L CO2 Incubator7 levels of shelving7
332342G01Shelving System to support G-Rex™ for Forma™ Steri-Cult™ 3310/3311 323L CO2 IncubatorForma Steri-Cult 3310/3311 CO2 Incubator3 shelves3
Número de catálogo 50164781
Precio (EUR)
3.390,00
Each
-
Añadir al carro de la compra
Descripción:
Shelving system to support up to 10 G-Rex 500M-CS bioreactors for Heracell VIOS 250i (CR), Forma Steri-Cycle i250 (CR) CO2 incubator
Para utilizar con (equipo):
Heracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series
Incluye:
3 expanded shelves with reinforced design, shelf supports and shelf rails
N.º de estantes:
3
Precio (EUR)
3.390,00
Each
Añadir al carro de la compra
Ask our AI about this Product
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space  
  • These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
  • With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
 Easy installation
  • Shelving systems are retrofittable and can be self-installed.
*G-Rex is a registered trademark of Wilson Wolf Corporation. G-Rex bioreactors must be ordered from Wilson Wolf Corporation.
Especificaciones
DescripciónShelving system to support up to 10 G-Rex 500M-CS bioreactors for Heracell VIOS 250i (CR), Forma Steri-Cycle i250 (CR) CO2 incubator
Para utilizar con (equipo)Heracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series
Incluye3 expanded shelves with reinforced design, shelf supports and shelf rails
MaterialAcero inoxidable (18/8)
N.º de estantes3
TipoAccesorio
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Compartir número de catálogo, nombre o enlace.
1x1 image pixel for data collection