Search Thermo Fisher Scientific
Description | Thermo Scientific OPC UA Gateway for Bio Safety Cabinets available in China only |
Product Type | Gateway Accessory Kit |
Includes | Power supply 100-240 V, USB 2.0 cable USB-A to USB-B connector |
For Use With (Equipment) | Herasafe™ 2030i, Maxisafe 2030i, Herasafe™ 2025, MSC-Advantage™, 1300 Series A2, and 1500 Series A2 Biological Safety Cabinets |
Voltage | 100/240 V |
Unit Size | Each |
Description | Thermo Scientific OPC UA Gateway for Bio Safety Cabinets available in all countries except for China |
Product Type | Gateway Accessory Kit |
Includes | Power supply 100-240 V, USB 2.0 cable USB-A to USB-B connector |
For Use With (Equipment) | Herasafe™ 2030i, Maxisafe 2030i, Herasafe™ 2025, MSC-Advantage™, 1300 Series A2, and 1500 Series A2 Biological Safety Cabinets |
Voltage | 100/240 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
50170005 | Each | Thermo Scientific OPC UA Gateway for Bio Safety Cabinets available in China only | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
50170006 | Each | Thermo Scientific OPC UA Gateway for Bio Safety Cabinets available in all countries except for China | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ OPC™ UA Gateway for Biological Safety Cabinets utilizes the OPC™ UA interoperability communication protocol to enable a secure and reliable exchange of real-time and historical data between Distributed Control Systems (DCS), such as the Emerson™ DeltaV™ system, and other systems, applications, and enterprise users.
What is OPC UA?
Open Platform Communication Unified Architecture (OPC™ UA) is a platform-independent, open industry standard for connectivity that enables machine-to-machine communication and information integration between various devices, software systems, and applications.
Advantages to connecting
The gateway accessory kit comes complete with a gateway device, USB 2.0 cable USB-A to USB-B connector, and 100-240 V power supply. One kit is needed per Biological Safety Cabinet connection.
The gateway accessory kit is compatible with the following models: