Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets (BSCs) combine safety and extraordinary value with energy efficiency, reliability and usability. With proprietary airflow design and energy-efficient DC motors, the MSC-Advantage BSCs use up to 68% less energy than cabinets with traditional AC motors. The cabinets provide both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. Cabinets are certified to EN 12469 safety standard.
Certifications/Compliance | EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) |
Width (Metric) Interior | 0.9 m |
Product Type | Biosafety Cabinet |
Noise Level | <55 dBA |
Receptacles | 1 Double, Right Side |
Width (English) Exterior | 39.3 in. |
Exhaust Air Filter | HEPA H 14 EN 1822, 99.995% at 0.3μm Particle |
Green Features | Energy efficient |
Width (Metric) Exterior | 100 cm |
Energy Consumption (Reduced Flow Mode) | <40 W |
Ports | TUV-approved |
Width (English) Interior | 2.95 ft. |
Height (Metric) Exterior | 152 cm |
Depth (Metric) Exterior | 80 cm |
Voltage | 230 V |
Energy Consumption (Operating Set Point) | 150 W |
Unit Size | Each |
Certifications/Compliance | EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) |
Width (Metric) Interior | 1.2 m |
Product Type | Biosafety Cabinet |
Noise Level | <59 dBA |
Receptacles | 1 Double, Right Side |
Width (English) Exterior | 51.1 in. |
Exhaust Air Filter | HEPA H 14 EN 1822, 99.995% at 0.3μm Particle |
Green Features | Energy efficient |
Width (Metric) Exterior | 130 cm |
Energy Consumption (Reduced Flow Mode) | <40 W |
Ports | TUV-approved |
Width (English) Interior | 4 ft. |
Height (Metric) Exterior | 152 cm |
Depth (Metric) Exterior | 80 cm |
Voltage | 230 V |
Energy Consumption (Operating Set Point) | 200 W |
Unit Size | Each |
Certifications/Compliance | EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) |
Width (Metric) Interior | 1.8 m |
Product Type | Biosafety Cabinet |
Noise Level | <59 dBA |
Receptacles | 2 Double, Left and Right Side |
Width (English) Exterior | 74.8 in. |
Exhaust Air Filter | HEPA H 14 EN 1822, 99.995% at 0.3μm Particle |
Green Features | Energy efficient |
Width (Metric) Exterior | 190 cm |
Energy Consumption (Reduced Flow Mode) | <70 W |
Ports | TUV-approved |
Width (English) Interior | 6 ft. |
Height (Metric) Exterior | 152 cm |
Depth (Metric) Exterior | 80 cm |
Voltage | 230 V |
Energy Consumption (Operating Set Point) | 340 W |
Unit Size | Each |
Certifications/Compliance | EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) |
Width (Metric) Interior | 1.5 m |
Product Type | Biosafety Cabinet |
Noise Level | <59 dBA |
Receptacles | 2 Double, Left and Right Side |
Width (English) Exterior | 62.9 in. |
Exhaust Air Filter | HEPA H 14 EN 1822, 99.995% at 0.3μm Particle |
Green Features | Energy efficient |
Width (Metric) Exterior | 160 cm |
Energy Consumption (Reduced Flow Mode) | <70 W |
Ports | TUV-approved |
Width (English) Interior | 4.92 ft. |
Height (Metric) Exterior | 152 cm |
Depth (Metric) Exterior | 80 cm |
Voltage | 230 V |
Energy Consumption (Operating Set Point) | 280 W |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Width (English) Interior | Width (Metric) Interior | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
51028225 | Each | 2.95 ft. | 0.9 m | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
51025411 | Each | 4 ft. | 1.2 m | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
51025413 | Each | 6 ft. | 1.8 m | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
51028226 | Each | 4.92 ft. | 1.5 m | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The MSC-Advantage biological safety is available in 4 sizes and can be connected to an external exhaust to provide protection for trace amounts of volatile toxic chemicals that are sometimes used in the cabinet.
Containment
Comfort
Convenience
Certifications/Compliance
Ordering Information:
Please order separately: Stand options, programmable UV light, one-piece work tray, ergonomic stool, free-standing floor shelf. Not available for sale in North America (equivalent model: 1300 Series A2 Biological Safety Cabinets).