Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next generation of mid-range Thermo Scientific™ Cytomat™ 2 C4xx-LiN series automated incubators for incubation/storage of cell culture applications.
These Cytomat incubators are available in various temperature ranges and gate positions for the hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec.
With a fully automated thermal decontamination andoptional controlled humidity, the Cytomat 2 C4xx-LiN series provides a new standard in significantly reducing the risk of contamination.
Certifications/Compliance | CE Marked |
Access Time | 12 sec. |
Chamber Material | Copper |
Holds | 56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well |
Relative Humidity | <90% |
Temperature Range (Metric) | Ambient +4° to 25°C |
Type | Automated Incubator |
Features | Incubation and storage unit with temperature range above; Humidity up to 90% r.H.; Includes decontamination routine-ContraCon |
For Use With (Application) | Incubation/storage for cell culture applications 4 to 25°C |
Model | Cytomat 2 C425-LiN 100V 50Hz |
Width (English) | 22.5 in. |
Width (Metric) | 572mm |
Height (English) | 35.6 in. |
Height (Metric) | 905mm |
Depth (English) | 20.1 in. |
Depth (Metric) | 511mm |
Weight (Metric) | 90kg |
Electrical Requirements | 100V 50Hz |
Voltage | 100 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Application) | Temperature Range (Metric) | Electrical Requirements | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
51033032 | Each | Incubation/storage for cell culture applications 4 to 25°C | Ambient +4° to 25°C | 100V 50Hz | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Cytomat 2 C4xx-LiN series of automated incubators provide an ideal safe environment for storage and incubation of assays. This series of mid-range incubators offer a choice of temperature ranges (4 to 70°C), high humidity up to 95% r.H. stable CO2 conditions, and automated ContraCon decontamination routine without harmful gases.
The external cooling and heating unit is designed to allow the user to create a customized working space for easy system integration into any automated assay system.
Available options