Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Accuracy | < 5% @ 300μL |
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 1 |
Bottles | Wash Bottle (2 L), Waste Bottle (2 L) |
Precision | < 3% (CV) @ 300μL (96-well plate) |
Dispense Volume | 50 to 400 μL |
Wash Heads | 1 x 12 |
Plate Type | 96-Well Plates |
Residual Volume | <2 μL |
Weight (Metric) | 8 kg |
Wash Volume | 50 to 1000 μL |
Display | LCD |
For Use With (Application) | ELISA-Based Assays |
Frequency | 50/60 Hz |
Model | Wellwash 1x12 |
Cell Washing | No |
Includes | 1 x 12 wash head, 1 x 2L wash bottle, 1 x 2L waste bottle, aerosol cover |
Width (English) | 15.2 in. |
Width (Metric) | 385 mm |
Height (English) | 13.6 in. |
Height (Metric) | 345 cm |
Depth (English) | 9.4 in. |
Depth (Metric) | 240 mm |
Automation Interface | No |
Weight (English) | 17.6 lb. |
Electrical Requirements | 100/240 V 50/60 Hz |
Voltage | 100/240 V |
Prime Volume | 5 to 100 mL |
Unit Size | Each |
Accuracy | < 5% @ 300μL |
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 1 |
Bottles | Wash Bottle (2 L), Waste Bottle (2 L) |
Precision | < 3% (CV) @ 300μL (96-well plate) |
Dispense Volume | 50 to 400 μL |
Wash Heads | 1 x 8 |
Plate Type | 96-Well Plates |
Residual Volume | <2 μL |
Weight (Metric) | 8 kg |
Wash Volume | 50 to 1000 μL |
Display | LCD |
For Use With (Application) | ELISA-Based Assays |
Frequency | 50/60 Hz |
Model | Wellwash 1x8 |
Cell Washing | No |
Includes | 1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, and aerosol cover. |
Width (English) | 15.2 in. |
Width (Metric) | 385 mm |
Height (English) | 13.6 in. |
Height (Metric) | 345 cm |
Depth (English) | 9.4 in. |
Depth (Metric) | 240 mm |
Automation Interface | No |
Weight (English) | 17.6 lb. |
Electrical Requirements | 100/240 V 50/60 Hz |
Voltage | 100/240 V |
Prime Volume | 5 to 100 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Model | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5165040 | Each | Wellwash 1x12 | 7,650.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5165000 | Each | Wellwash 1x8 | 7,650.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Includes:
1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover
Recommended for:
ELISA-based assays
Warranty: 1 year