Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Accuracy | < 5% @ 300μL, with n x 8 or n x 12 wash head; < 5% @ 250μL, with 2 x 8 cell wash head; < 5% @ 100μL, with 1 x 16 wash head |
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 3 |
Bottles | Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) |
Precision | < 3% (CV) @ 300μL, with n x 8 or n x 12 wash heads |
Dispense Volume | 50 to 400 μL (96); 20 to 120 μL (384) |
Wash Heads | Up to two 1 x 12 wash heads and optional up to two 1 x 8 wash heads and 2 x 8 cell wash head for 96 well plate; 1 x 16 wash head for 384 well plate |
Plate Type | 96-well Plate, 384-well Plate |
Residual Volume | <2 μL |
Weight (Metric) | 9 kg |
Wash Volume | 50 to 1000 μL (96); 20 to 300 μL (384) |
Rinse Volume | 5 to 100 mL |
Display | LCD |
For Use With (Application) | ELISA-Based Assays; Cell Washing |
Frequency | 50/60 Hz |
Model | Wellwash Versa 2x12 |
Cell Washing | Yes |
Includes | 2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 2x12 wash heads, and aerosol cover |
Width (English) | 15.2 in. |
Width (Metric) | 385 mm |
Height (English) | 13.6 in. |
Height (Metric) | 345 mm |
Depth (English) | 9.4 in. |
Depth (Metric) | 240 mm |
Automation Interface | Yes (USB or RS-232C) |
Weight (English) | 20 lb. |
Electrical Requirements | 100/240 V 50/60 Hz |
Voltage | 100/240 V |
Prime Volume | 5 to 100 mL |
Unit Size | Each |
Accuracy | < 5% @ 300μL, with n x 8 or n x 12 wash head; < 5% @ 250μL, with 2 x 8 cell wash head; < 5% @ 100μL, with 1 x 16 wash head |
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 3 |
Bottles | Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) |
Precision | < 3% (CV) @ 300μL, with n x 8 or n x 12 wash heads |
Dispense Volume | 50 to 400 μL (96); 20 to 120 μL (384) |
Wash Heads | Up to two 1 x 8 wash heads and optional up to two 1 x 12 wash heads and 2 x 8 cell wash head for 96 well plate; 1 x 16 wash head for 384 well plate |
Plate Type | 96-well Plate, 384-well Plate |
Residual Volume | <2 μL |
Weight (Metric) | 9 kg |
Wash Volume | 50 to 1000 μL (96); 20 to 300 μL (384) |
Rinse Volume | 5 to 100 mL |
Display | LCD |
For Use With (Application) | ELISA-Based Assays; Cell Washing |
Frequency | 50/60 Hz |
Model | Wellwash Versa 2x8 |
Cell Washing | Yes |
Includes | 2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 1x8 wash heads, and aerosol cover. |
Width (English) | 15.2 in. |
Width (Metric) | 385 mm |
Height (English) | 13.6 in. |
Height (Metric) | 345 mm |
Depth (English) | 9.4 in. |
Depth (Metric) | 240 mm |
Automation Interface | Yes (USB or RS-232C) |
Weight (English) | 20 lb. |
Electrical Requirements | 100/240 V 50/60 Hz |
Voltage | 100/240 V |
Prime Volume | 5 to 100 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Model | Price (KRW) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5165050 | Each | Wellwash Versa 2x12 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5165010 | Each | Wellwash Versa 2x8 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Easy and convenient to use
Reliable and secure performance
Optimal washing performance
Added versatility
Includes:
2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 1x8 wash heads, aerosol cover
Recommended for:
ELISA-based assays, cell washing
Warranty: 1 year