Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 490/525 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 488 |
Color | Green |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 401/421 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 405 |
Color | Blue |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; and Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 590/617 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 594 |
Color | Red |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; and Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 784/814 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 800 |
Color | Near-IR |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; and Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 562/576 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 550 |
Color | Yellow |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; and Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 652/672 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 650 |
Color | Far-Red |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Kit Contents | Contains DyLight Fluor NHS Ester, 5 x 15 μg vials; Borate Buffer (0.67 M), 1 mL; Purification Resin, 5 mL; Microcentrifuge Spin Columns, 5 columns; and Microcentrifuge Collection Tubes, 10 Tubes |
Product Type | Labeling Kit |
Excitation/Emission | 749/775 |
Labeling Scale | 100 μg |
Product Line | DyLight™ |
Chemical Reactivity | Amine |
Label or Dye | DyLight 755 |
Color | Near-IR |
Labeling Target | Antibodies, Proteins |
Label Type | DyLight™ |
Unit Size | 5 reactions |
Catalog Number | Specifications | Unit Size | Excitation/Emission | Label or Dye | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
53025 | 5 reactions | 490/525 | DyLight 488 | 499.65 Online price 526.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
53021 | 5 reactions | 401/421 | DyLight 405 | 497.65 Online price 524.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
53045 | 5 reactions | 590/617 | DyLight 594 | 494.65 Online price 520.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
53063 | 5 reactions | 784/814 | DyLight 800 | 494.65 Online price 520.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
84531 | 5 reactions | 562/576 | DyLight 550 | 495.65 Online price 522.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
84536 | 5 reactions | 652/672 | DyLight 650 | 498.65 Online price 526.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
84539 | 5 reactions | 749/775 | DyLight 755 | 495.65 Online price 522.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|