Richard-Allan Scientific™ Bio-Tite™ Specimen Containers, 360mL
Richard-Allan Scientific™ Bio-Tite™ Specimen Containers, 360mL
Thermo Scientific™
Richard-Allan Scientific™ Bio-Tite™ Specimen Containers, 360mL
Transport, store and retrieve your pathology specimens in a safe and cost-effective manner with Thermo Scientific™ Richard-Allan Scientific™ Bio-Tite™ Specimen Containers.
Have Questions?
Catalog number 5312
Price (USD)
-
Request A Quote
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™). This system includes containers pre-filled with the highest quality 10% Neutral Buffered Formalin (NBF) and Zinc Formalin.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Thermo Scientific™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Thermo Scientific Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Closure IncludedYes
MaterialPolyethylene
SterilitySterile
DescriptionDry Containers
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link