Pierce™ NeutrAvidin™ UltraLink™ Resin
Pierce™ NeutrAvidin™ UltraLink™ Resin
Thermo Scientific™

Pierce™ NeutrAvidin™ UltraLink™ Resin

Thermo Scientific NeutrAvidin UltraLink Resin is a beaded polyacrylamide resin of immobilized NeutrAvidin Protein, a modified form of avidin withRead more
Have Questions?
Catalog NumberQuantity
531505 mL
Catalog number 53150
Price (USD)
478.00
Each
In stock
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)
478.00
Each
Add to cart
Thermo Scientific NeutrAvidin UltraLink Resin is a beaded polyacrylamide resin of immobilized NeutrAvidin Protein, a modified form of avidin with exceptional biotin-binding characteristics for affinity purification methods.

Features of NeutrAvidin UltraLink Resin:

NeutrAvidin protein—purified, deglycosylated avidin protein (60kDa, pI 6.3); tetrameric with four biotin-binding site per molecule
UltraLink Resin—acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium-pressure applications
Standard capacity—this variety of beads has a normal load of immobilized NeutrAvidin protein, providing a binding capacity of 1 to 2 mg of biotinylated BSA per mL of resin

NeutrAvidin UltraLink Resin consists of a specially purified and deglycosylated form of avidin that has been covalently immobilized onto a beaded acrylamide support. Compared to native avidin, NeutrAvidin protein is more neutral and exhibits much less nonspecific binding properties in biotin-binding applications. UltraLink beads are hydrophilic, charge-free, highly crosslinked, rigid and durable. These properties make the affinity resin suitable for a variety of batch- or column-type affinity procedures that require medium-pressure chromatography. The resin provides an alternative to agarose supports when situations demand it. Generally, NeutrAvidin products function as well or better than their streptavidin counterparts and are less expensive.

NeutrAvidin protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. NeutrAvidin resins are prepared by covalently coupling the protein using efficient and stable chemistries, resulting in supports that are resistant to leaching and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Properties of Thermo Scientific UltraLink Resin:
• Support pH Stability: 1 to 13
• Average Particle Size: 50 to 80 microns
• Exclusion Limit: greater than 2,000,000 daltons
• Average Surface Area: greater than 250m2 per gram of beads
• Average Pore Volume: greater than 1.2 mL per gram of beads (>60% of bead volume)
• Pore Size: 500 to 1000 angstroms (50 to 100nm)
• Maximum Linear Velocity: 3000 cm per hour
• Maximum Pressure: 100psi (6.9 bar)

Related Products
Pierce™ NeutrAvidin™ Agarose
Pierce™ High Capacity NeutrAvidin™ Agarose
Pierce™ NeutrAvidin™ Plus UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, UltraLink™ Resin
DescriptionPierce NeutrAvidin UltraLink Resin
FormatBottle
Quantity5 mL
Stationary PhaseNeutrAvidin Protein
TargetBiotin-Tagged Molecule
Capacity (Metric)1 to 2 mg Biotinylated BSA Binding per mL Resin
Particle Size50 to 80 μm
Product LineNeutrAvidin™, Pierce™, UltraLink™
TypeResin
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Figures

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
ZI376083Certificate of AnalysisOct 15, 202453150
YG361885Certificate of AnalysisAug 10, 202353150
XI361884Certificate of AnalysisSep 24, 202253150
XF349495Certificate of AnalysisJun 20, 202253150
XB339995Certificate of AnalysisFeb 12, 202253150
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection