Storage Tube Protection
Storage Tube Protection
Storage Tube Protection
Storage Tube Protection
Storage Tube Protection
Thermo Scientific™
Storage Tube Protection
Give your cryogenic storage tubes and vials extra protection, such as when shipping or storing in liquid or vapor phase nitrogen with these storage accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
534592Cryo Storage Box200/Pk., 200/Cs.
343958Tube Wrap15/Pk., 300/Cs.
343958PKTube Wrap15/Pk., 300/Cs.
5016-0001Vial Holder100/Pk., 100/Cs.
534479Cryo Storage Box350/Pk., 350/Cs.
Catalog number 534592
Price (USD)
846.00
Online Exclusive
987.00
Save 141.00 (14%)
Case of 200
In stock
Add to cart
Type:
Cryo Storage Box
Quantity:
200/Pk., 200/Cs.
Price (USD)
846.00
Online Exclusive
987.00
Save 141.00 (14%)
Case of 200
Add to cart
Give your cryogenic storage tubes and vials extra protection when needed with Thermo Scientific™ Storage Tube Protection accessories. For example, safeguard your samples when storing in liquid or vapor phase nitrogen, or when shipping samples. These cryogenic storage accessories, such as tube wraps, sleeves and mini-boxes, have safe handling in mind.
  • If you want to submerge your cryotubes in the liquid phase of nitrogen, seal them first inside Nunc™ Cryoflex™ tubing to prevent liquid nitrogen from infiltrating the tube. Cryoflex™ tubing can used to crimp and heat shrink an impenetrable safety skin around the tube prior to submersion.
  • Nalgene™ CryoSleeves™ slide over any Nunc or Nalgene Cryocane and prevent tubes from falling out of the cryocane and into the bottom of the liquid nitrogen dewar. Nalgene™ CryoSleeves™ do not prevent infiltration of liquid nitrogen. Cryogenic tubes mounted in the cane must first be sealed inside Cryoflex™ tubing prior to applying the CryoSleeve™ if liquid phase submersion is required.
  • Nunc CryoTube™ miniboxes with a friction-fit lid are ideal for storing small quantities of samples and shipping.Made with durable, high-impact, polystyrene to protect samples.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
DescriptionCryoTube Mini Box, For 10 vials
ColorTan
MaterialPolystyrene
Quantity200/Pk., 200/Cs.
TypeCryo Storage Box
Unit SizeCase of 200
nalgene-certificate-of-compliance
Certificates of Compliance
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link