KingFisher™ Purification System
This SKU is pending discontinuation.
KingFisher™ Purification System
Thermo Scientific™

KingFisher™ Purification System

Automate time-consuming sample preparation processes of nucleic acids, proteins and cells with Thermo Scientific™ KingFisher™ Purification Systems.
Have Questions?
Catalog NumberQuantity
5400000Each
Catalog number 5400000
Price (USD)
-
Request A Quote
Quantity:
Each

The KingFisher™ Purification System is pending discontinuation. Thermo Fisher Scientific offers other automation platforms such as the KingFisher Duo Prime Purification System, KingFisher Flex Purification System and KingFisher Apex Purification System. The company will continue to support current KingFisher Purification System users with plastics, service, and technical support through 2028, or best effort.

By August 1st, 2023, Thermo Fisher Scientific will transition to limited support stage. It will offer limited support contracts and provide best-effort service and support, subject to availability of spare parts and resources.

If you are interested in upgrading to one of its newer sample preparation instruments, please contact your local account manager for product information.

Features:
  • Offer rapid and reproducible purification of high-quality DNA, RNA, proteins and cells for various types of downstream applications
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification
  • Ready-made protocols for different types of applications available

Recommended for:

Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays

For research use only. Not for use in diagnostic procedures.
Specifications
Collection Efficiency of Particles99%
Depth (English)11.4 in.
Depth (Metric)29 cm
DescriptionKingfisher Purification System, 100-240V, 50/60Hz
DisplaySTART/STOP/TWO CURSOR KEYS/LCD
Electrical Requirements100-240 V, 50/60 Hz
For Use With (Application)Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays
Height (English)12.2 in.
Height (Metric)31 cm
Magnetic Rods2 x 12 format
Particle Size (Metric)ca. >1 μm
Plate TypeSpecial design, 8 x 12 grid format
QuantityEach
Tip ComboSpecial design, 2 x 12 format
Weight (English)23 lb.
Weight (Metric)10 kg
Width (English)11.4 in.
Width (Metric)29 cm
Capacity24 Samples/Run
Volume (Metric) Processing20 to 200 μL
Unit SizeEach
Please enter sku to load default questions

Frequently asked questions (FAQs)

What is BindIt Software?

BindIt Software is used to operate the KingFisher 96, KingFisher mL, KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto instruments. With the software you can:
• Create a plate layout
• Create new or modify existing protocols
• Transfer protocols to the KingFisher instrument memory
• Delete protocols from the KingFisher instrument
• Execute a protocol directly from the software without first sending it to the KingFisher instrument memory
• Export protocols to other databases
• Import protocols to the PC
• View protocol status reports

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link