Thermo Scientific™

KingFisher™ mL Food Protection Purification System

Catalog number: 5400050C
Have Questions?
Thermo Scientific™

KingFisher™ mL Food Protection Purification System

Catalog number: 5400050C
Have Questions?
Catalog Number
5400050C
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
5400050CEach
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Carry out rapid purification and concentration of selected targets from a broad range of pre-enriched food samples with the Thermo Scientific KingFisher mL Food Protection Purification System. Combined with Dynabeads immuno-magnetic separation (IMS) products, the system provides an optimized solution for the purification of microorganisms prior to detection using methods such as PCR, ELISA, and agar plates.

Features of the KingFisher mL Food Protection Purification System include:
• Protocols included for E. Coli 0157, EPEC/VTEC, listeria, and salmonella
• Automated processing of up to 15 samples in ∼40 min
• High reproducibility and maximum performance of capture technology
• Simple, easy operation: complete processing through internal program
• Small footprint: uses only 11.8 in (30 cm) of bench depth

The system-of-choice for processing volumes up to 1 mL, this magnetic particle processor enables all purification steps in a single-strip of five tubes, and supports 15 processed samples per run. With the ability to release target molecules into a volume as small as 50 μL, samples such as DNA or RNA can be isolated and concentrated from larger starting volumes simultaneously.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
KingFisher Purification System
Sample Type
Food
Product Line
KingFisher™
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text