Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Volume (Metric) Processing | 20 to 100 μL |
Plates per Deck | 24 or 96 |
Format | 96-well Plate |
Capacity | 96 Samples |
Description | KingFisher 96 plate, PCR plate |
Collection Efficiency of Particles | > 95% |
Display | START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD |
For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics |
Heating Temperature | From +5°C above ambient temperature to +115°C |
Magnetic Rods | 96 in one frame; Interchangeable magnet heads |
Particle Size (Metric) | ca. >1 μm |
Plate Type | 24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL) |
Depth (English) | 23.6 in. |
Depth (Metric) | 60 cm |
Height (English) | 15 in. |
Height (Metric) | 38 cm |
Weight (English) | 62 lb. |
Weight (Metric) | 28 kg |
Width (English) | 26.8 in. |
Width (Metric) | 68 cm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5400610 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Recommended for: