KingFisher™ Flex Purification System, KingFisher with 96 Deep-well Head
Inquire about OEM or Commercial Supply version of this product here.
KingFisher™ Flex Purification System, KingFisher with 96 Deep-well Head
Thermo Scientific™

KingFisher™ Flex Purification System, KingFisher with 96 Deep-well Head

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processors.
Have Questions?
Catalog number 5400630
Price (USD)/ Each
-
Request A Quote
Prepare samples efficiently from a variety of materials with the Thermo Scientific™ KingFisher™ Flex Purification System, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.
Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20 minutes.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.

Recommended for:

  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics

For Laboratory Use
Specifications
Volume (Metric) Processing50 to 1000 μL
Plates per Deck24 or 96
Capacity96 Samples
DescriptionKingFisher 96 Plate, KingFisher 96 DW plate
Collection Efficiency of Particles> 95%
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating TemperatureFrom +5°C above ambient temperature to +115°C
Magnetic Rods96 in one frame; Interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
QuantityEach
Plate Type24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
Depth (English)23.6 in.
Depth (Metric)60 cm
Height (English)15 in.
Height (Metric)38 cm
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Unit SizeEach