Search Thermo Fisher Scientific
Catalog Number | Product Type |
---|---|
5400910 | 96 PCR Head |
A46941 | 96 PCR head, Ext Warranty |
5400920 | 96 Combi head |
A46943 | 96 Combi head, Ext Warranty |
5400930 | 96 DW head |
A46944 | 96 DW head, Ext Warranty |
5400940 | 24 Combi head |
A46942 | 24 Combi Head, Ext Warranty |
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
Share catalog number, name or link