Search Thermo Fisher Scientific
Description | PCR 630 online rheometer |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
550-0030 | Each | PCR 630 online rheometer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
All HAAKE PCR systems have independent motor control for each pump, which allows fast and flexible control of the systems. The HAAKE PCR 620 is a 2 pump system giving fast response for products with medium and high melt index. The HAAKE PCR 620 is a 3 pump bypass system which allows even short response time for product grades with low melt index.
Recommended for:
Applications:
Materials: