Axiom™ Bovine-Ovine-Caprine Genotyping Array
Axiom™ Bovine-Ovine-Caprine Genotyping Array
Applied Biosystems™

Axiom™ Bovine-Ovine-Caprine Genotyping Array

Axiom Bovine-Ovine-Caprine Genotyping Array (Axiom Ovicap Array) for animal genotyping was designed through the Affymetrix Expert Design Program. The arrayAfficher plus
Have Questions?
RéférenceQuantité
5506271 plate
Référence 550627
Prix (EUR)
-
Quantité:
1 plate
Axiom Bovine-Ovine-Caprine Genotyping Array (Axiom Ovicap Array) for animal genotyping was designed through the Affymetrix Expert Design Program. The array includes single nucleotide polymorphisms (SNPs) that were identified by the United States Department of Agriculture Agricultural Research Service (USDA ARS), International Sheep Genomics Consortium (ISGC), and the International Goat Genome Consortium for each of the three different species (bovine, ovine, and caprine, respectively. The content for caprine includes SNPs located within the four casein genes implicated in milk production.The array also includes markers for ovine and bovine parentage analysis.

Highlights
• Expert design: developed by key opinion leaders in the community
• Informative, including parentage markers from existing in-market arrays, allowing compatibility with previous studies
• A singe array for three species:
   - Bovine: 54,560 markers.
   - Ovine: 54,236 markers.
   - Caprine: 60,034 markers.

Applications
• Construction of high-resolution genetic maps
• Genetic improvement of pure lines
• Fine mapping of quantitative trait loci (QTL)
• Calculating breeding values
• Parentage analysis

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument

Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic.

Spécifications
À utiliser avec (application)Analyse de microarrays
Quantité1 plate
ArrayGenotyping
Format96-array Plate
Nombre de matrices96 matrices
EspècesBovine, Caprine, Ovine
Unit Size1 plate

Partager la référence catalogue, le nom ou le lien.