Thermo Scientific™

PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Catalog number: 5756-250
Have Questions?
Thermo Scientific™

PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Catalog number: 5756-250
Have Questions?

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.

 
Catalog Number
5756-250
Unit Size
Case of 7680
Volume (Metric)
250 nL
Sterility
Non-sterile
Packaging
20 x 384 tips
Price (USD)
Full specifications
Volume (Metric)250 nL
FilteredNon-Filtered
SterilityNon-sterile
FormatRack
Tip StyleAutomation
Packaging20 x 384 tips
DisposableSingle-use
For Use With (Equipment)Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product LinePocketTip, PocketTip
Unit SizeCase of 7680
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeVolume (Metric)SterilityPackagingPrice (USD)
5756-250Full specifications
Case of 7680250 nLNon-sterile20 x 384 tipsRequest A Quote
Volume (Metric)250 nL
FilteredNon-Filtered
SterilityNon-sterile
FormatRack
Tip StyleAutomation
Packaging20 x 384 tips
DisposableSingle-use
For Use With (Equipment)Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product LinePocketTip, PocketTip
Unit SizeCase of 7680
Showing 1 of 1

Features:

  • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
  • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
  • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
  • Control consistent accuracy and precision during the molding process
Validation and Compatibility:
  • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
  • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
  • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
  • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
Ordering Information:
  • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
  • Sterile format available for sensitive liquid handling procedures
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text