Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Maintain protein integrity and labile post-translational modifications during separation at standard analytical flow rates. The Thermo Scientific™ Dionex™ UltiMate™ WPS TBPL Thermostatted Biocompatible Standard Well Plate Autosampler features a biocompatible flow path consisting only of fused silica, Thermo Scientific™ PEEK™ and high-performance polymers. Pulled-loop injection mechanism reduces delay times, provides accurate and precise gradient profiles, reduces peak broadening and improves peak resolution. This biocompatible autosampler extends these capabilities to pressures up to 345 bar.
Type | Analytical Biocompatible Pulled-Loop Thermostatted Well Plate Autosampler |
Injection Cycle Time | Full Loop <60; Partial Loop <90 sec. |
Injection Volume Precision | <0.25% |
Injection Volume Range | 0.01 to 250 μL |
Injection Type | Pulled Loop |
Sample Capacity | 3 Racks or Well Plates |
Max. Pressure | 345 bar |
Biocompatible Version | Yes |
Linearity | >0.9999 |
Voltage | 100 to 120 V |
Weight (Metric) | 23 kg |
Description | WPS-3000 TBPL Analytical Biocompatible Pulled-Loop Thermostatted Well Plate Autosampler for analytical flow rates |
Carryover | <0.02% |
For Use With (Equipment) | UltiMate 3000 Series LC Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Sample Capacity | Max. Pressure | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5823.0020 | Each | WPS-3000 TBPL Analytical Biocompatible Pulled-Loop Thermostatted Well Plate Autosampler for analytical flow rates | UltiMate 3000 Series LC Systems | 3 Racks or Well Plates | 345 bar | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Recommended Use
The biocompatible WPS TBPL Thermostatted Biocompatible Standard Well Plate Autosampler is an ideal tool for biomolecule analysis, including protein and antibody fractionation and purification. The stainless-steel version can be upgraded to support up to 90MPa (13,000 psi) for full support of fraction collection at UHPLC conditions. In combination with the extended fraction collection capabilities of Thermo Scientific™ Dionex™ Chromeleon™ Chromatography Data System (CDS) software, the WPS TBPL enables full automation of injection, fractionation and reinjection of samples.
Performance and Features