Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
Accuracy | Small tube dispensing cassette: 2μL, ±10%; 10μL, ±5% Standard tube dispensing cassette: 20μL, ±2%; 100μL, ±1% |
Product Type | Reagent Dispenser |
Description | Multidrop™ Combi+ Reagent Dispenser |
Channels | 8 |
Volume (Metric) | 0.5 to 2500 μL |
For Use With (Application) | The instrument is intended for automated dispensing of up to eight different reagents simultaneously into 6, 12, 24, 48, 96, or 384-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm. |
For Use With (Equipment) | Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and a variety of microplates and strips in 96, 384 or 1536-well plate format |
Includes | One standard tube dispensing cassette (cat. no. 24072670) |
Interface | On-board software and Serial RS-232 or USB for PC control |
Plate Type | 6-1536 - 6, 12, 24, 48, 96, and 384 well plates |
Precision |
|
Speed |
|
Volume (English) |
|
Volume (Metric) Dead | <1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent) |
Warranty | 1 year |
Dimensions (L x W x H) | 14 x 8.6 x 12.9 in.; 35.5 x 22 x 33 cm |
Weight (English) | 20.1 lb |
Weight (Metric) | 9.1 kg |
Unit Size | Each |
Accuracy |
|
Product Type | Reagent Dispenser |
Description | Multidrop™ Combi SMART+ Reagent Dispenser |
Channels | 8 |
For Use With (Application) | The instrument is intended for automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. |
For Use With (Equipment) | Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and 6, 12, 24, 48, 96, or 384-well plate format. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm. |
Includes | One SMART+ standard tube dispensing cassette (cat. no. N22704) |
Interface | On-board software and Serial RS-232 or USB for PC control |
Plate Type | 6, 12, 24, 48, 96, and 384 well plates |
Precision |
|
Speed |
|
Volume (English) |
|
Volume (Metric) Dead | <1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent) |
Warranty | 1 year |
Dimensions (L x W x H) | 14 x 8.6 x 12.9 in. (35.5 x 22 x 33 cm) |
Weight (English) | 20.1 lb. |
Weight (Metric) | 9.1 kg |
Electrical Requirements | 100 to 240 Vac 50/60Hz |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5840330 | Each | Multidrop™ Combi+ Reagent Dispenser | 20,722.50 Online price 23,025.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5840340 | Each | Multidrop™ Combi SMART+ Reagent Dispenser | 25,376.40 Online price 28,196.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
Multidrop Combi SMART:
Recommended for: