Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 45mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 1000 mL |
Diameter (Metric) | 90 mm |
Diameter (Metric) Membrane | 90 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 33mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 150 mL |
Diameter (Metric) | 50 mm |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 45mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 150 mL |
Diameter (Metric) | 50 mm |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 33mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 150 mL |
Diameter (Metric) | 50 mm |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 45mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 150 mL |
Diameter (Metric) | 50 mm |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 33mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 500 mL |
Diameter (Metric) | 75 mm |
Diameter (Metric) Membrane | 75 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 45mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 500 mL |
Diameter (Metric) | 75 mm |
Diameter (Metric) Membrane | 75 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 33mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 500 mL |
Diameter (Metric) | 75 mm |
Diameter (Metric) Membrane | 75 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 45mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 500 mL |
Diameter (Metric) | 75 mm |
Diameter (Metric) Membrane | 75 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Filter Type | Membrane Filter |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Fits Bottle Neck Size (Metric) | 33mm |
No. per Case | 12 |
No. per Pack | 12 |
Type | Bottle Top Filter |
Volume (Metric) | 1000 mL |
Diameter (Metric) | 90 mm |
Diameter (Metric) Membrane | 90 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Catalog Number | Specifications | Unit Size | Material (Membrane) | Volume (Metric) | Pore Size | Fits Bottle Neck Size (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
597-4520 | Case of 12 | PES | 1000 mL | 0.2 μm | 45mm | 199.20 Online price 249.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
596-3320 | Case of 12 | PES | 150 mL | 0.2 μm | 33mm | 114.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
596-4520 | Case of 12 | PES | 150 mL | 0.2 μm | 45mm | 90.40 Online price 113.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
296-3345 | Case of 12 | PES | 150 mL | 0.45 μm | 33mm | 120.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
296-4545 | Case of 12 | PES | 150 mL | 0.45 μm | 45mm | 115.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
595-3320 | Case of 12 | PES | 500 mL | 0.2 μm | 33mm | 146.65 Online price 156.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
595-4520 | Case of 12 | PES | 500 mL | 0.2 μm | 45mm | 124.80 Online price 156.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
295-3345 | Case of 12 | PES | 500 mL | 0.45 μm | 33mm | 140.40 Online price 156.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
295-4545 | Case of 12 | PES | 500 mL | 0.45 μm | 45mm | 140.40 Online price 156.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
597-3320 | Case of 12 | PES | 1000 mL | 0.2 μm | 33mm | 252.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|