HyperSep™ Universal Vacuum Manifold
HyperSep™ Universal Vacuum Manifold
Thermo Scientific™
HyperSep™ Universal Vacuum Manifold
Use this flexible vacuum manifold to process samples using both solid phase extraction (SPE) cartridges and 96 well plates.
Have Questions?
Catalog NumberDescriptionFor Use With (Application)
60104-230Universal Vacuum Manifold, 1 pkSPE
Catalog number 60104-230
Price (USD)
3,784.73
Each
-
Add to cart
Description:
Universal Vacuum Manifold, 1 pk
For Use With (Application):
SPE
Recurring order eligible. Learn more »
Price (USD)
3,784.73
Each
Add to cart
Ask our AI about this Product
Process your samples in solid phase extraction SPE cartridges or 96 well plates by using the flexible Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. The manifold is supplied with a base and vacuum gauge, flask and stopper tubing and spigots.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionUniversal Vacuum Manifold, 1 pk
For Use With (Application)SPE
For Use With (Equipment)SPE Columns and 96 Well Plates
Product LineHyperSep
TypeUniversal Vacuum Manifold
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
The Pluripotent Stem Cell Immunocytochemistry Kit provides the NucBlue Fixed Cell Stain. If I don't have the correct filter set for this stain, can I use another nucleic acid stain?
Do you offer the individual components of the Pluripotent Stem Cell Immunocytochemistry Kit as standalone products?
Can I use the Pluripotent Stem Cell Immunocytochemistry Kit for flow cytometry analysis?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Videos
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Kunden, die diesen Artikel ansahen, interessierten sich auch für
      Documents & Downloads
      Certificates
      0 results displayed, search above for a specific certificate
      Share catalog number, name or link
      1x1 image pixel for data collection