HyperSep™ C8 Cartridges
HyperSep™ C8 Cartridges
Thermo Scientific™

HyperSep™ C8 Cartridges

Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 cartridges offer excellent performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightColumn Capacity (Metric)
60108-309500 mg3 mL
60108-39150 mg1 mL
60108-392100 mg1 mL
60108-393200 mg3 mL
60108-394500 mg6 mL
60108-4271000 mg6 mL
60108-7042000 mg15 mL
60108-70610 g75 mL
Catalog number 60108-309
Price (USD)
191.69
Pack of 50
Add to cart
Bed Weight:
500 mg
Column Capacity (Metric):
3 mL
Price (USD)
191.69
Pack of 50
Add to cart
Ask our AI about this Product

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ C8 Cartridges offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:

  • Drugs and their metabolites in biological samples
  • Peptides in biological samples
HyperSep C8 SPE Cartridges provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity (Metric)3 mL
Description500 mg Bed Weight; 3 mL Column Volume
For Use With (Application)Screening of Drugs, Metabolites, Peptides in Biological Samples
QuantityPack of 50
Bed Weight500 mg
For Use With (Equipment)Thermo Scientific HyperSep-96 System
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C8
TypeSolid Phase Extraction Cartridge
Volume (Metric)3 mL
Unit SizePack of 50
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection