Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
Catalog Number
60114-103
Unit Size
Each
Description
SMART Digest Bulk Protein A Fractionation Kit, Magnetic, with SOLAμ Collection Plate, Soluble
Price (USD)
1,842.36
Full specifications
Quantity
Description
SMART Digest Bulk Protein A Fractionation Kit, Magnetic, with SOLAμ Collection Plate, Soluble
Capture
Protein A
Cleavage
C-terminal side of lysine and arginine amino acids
Enzyme
Trypsin
Format
SMART Digest Soluble Resin, Bulk Magnetic
For Use With (Application)
Immunocapture and Protein Digest Sample Preparation
SMART Digest Bulk Protein A Fractionation Kit, Magnetic, Standalone
783.95
Description
SMART Digest Bulk Protein A Fractionation Kit, Magnetic, Standalone
Capture
Protein A
Enzyme
None
Format
SMART Digest Resin, Bulk Magnetic
For Use With (Application)
Immunocapture and Sample Preparation
Product Line
SMART Digest
Type
Immunocapture
Includes
Bulk Magnetic SMART Digest Resin
Unit Size
Each
Showing 5 of 5
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.