Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
Catalog Number
60115-104
Unit Size
Each
Description
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Soluble
Price (USD)
1,546.55
Full specifications
Quantity
Description
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Soluble
Capture
Protein G
Cleavage
C-terminal side of lysine and arginine amino acids
Enzyme
Trypsin
Format
SMART Digest Soluble Resin, Bulk Magnetic
For Use With (Application)
Immunocapture and Protein Digest Sample Preparation
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Standalone
783.95
Description
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Standalone
Capture
Protein G
Enzyme
None
Format
SMART Digest Resin, Bulk Magnetic
For Use With (Application)
Immunocapture and Sample Preparation
Product Line
SMART Digest
Type
Immunocapture
Includes
Bulk Magnetic SMART Digest Resin
Unit Size
Each
Showing 5 of 5
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.