SMART Digest™ Bulk Fractionation Kits: Streptavidin
SMART Digest™ Bulk Fractionation Kits: Streptavidin
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Streptavidin
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60119-104SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin
60118-101SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, without Soluble Trypsin
60119-103SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
Catalog number 60119-104
Price (USD)
1,724.31
Each
Add to cart
Description:
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin
Price (USD)
1,724.31
Each
Add to cart
Ask our AI about this Product
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
DescriptionSMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin
CaptureStreptavidin
CleavageC-terminal side of lysine and arginine amino acids
EnzymeTrypsin
FormatSMART Digest Resin, Bulk Magnetic
For Use With (Application)Immunocapture and Protein Digest Sample Preparation
Product LineSMART Digest
TypeImmunocapture + Protein Digestion (separate)
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Soluble Trypsin
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link